Anti-GP111 Rabbit Polyclonal Antibody
Catalog # ABGEAP12583C
Supplier: Abgent
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:G Protein-Coupled Receptor 111
- Antigensymbol:GPR111
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:middle
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:400 µl
Specifications
About this item
Type: Primary
Antigen: GP111
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human