Order Entry
Sweden
ContactUsLinkComponent
Anti-DPH7 Rabbit Polyclonal Antibody
Anti-DPH7 Rabbit Polyclonal Antibody
  ABGEAP12477A
 :  Abgent
Anti-DPH7 Rabbit Polyclonal Antibody
  ABGEAP12477A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    WD Repeat Domain 85
  • Antigen symbol:
    WDR85
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    FSHD region gene 2 protein family member B|FRG2B_HUMAN|HSA10-FRG2|Protein FRG2-like-1.|FRG2B|FRG2C|FRG2A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: DPH7 (diphthamide biosynthesis 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human