Anti-EVPLL Rabbit Polyclonal Antibody
Catalog # ABGEAP12312C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 35 Member G5
- Antigen symbol:EVPLL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:AMAC|Acyl-Malonyl-Condensing Enzyme 1-Like Protein 2|AMAC1L2|Protein AMAC1L2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: EVPLL (envoplakin-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human