Anti-SRC Rabbit Polyclonal Antibody
Catalog # ABGEAP12262C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor coactivator 1
- Antigen symbol:NCOA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:MGC129720|bHLHe74|KAT13A|RIP160|NCOA1|6/G9a|SRC1|bHLHe42|NCoA¡1|SRC¡1|F¡SRC¡1|MGC129719
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human