Anti-REG3A Rabbit Polyclonal Antibody
ABGEAP11691C
: Abgent
- Antibody type:Primary
- Antigen name:Poly(A) polymerase alpha
- Antigen symbol:PAPOLA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:ROBLD2|DNLC2B|DNCL2B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: REG3A (regenerating islet-derived 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human