Order Entry
Sweden
ContactUsLinkComponent
Anti-POLR3GL Rabbit Polyclonal Antibody [clone: 208CT34.1.1]
Anti-POLR3GL Rabbit Polyclonal Antibody [clone: 208CT34.1.1]
  ABGEAP11367C
 :  Abgent
Anti-POLR3GL Rabbit Polyclonal Antibody [clone: 208CT34.1.1]
  ABGEAP11367C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 622
  • Antigen symbol:
    POLR3GL
  • Clonality:
    Polyclonal
  • Clone:
    208CT34.1.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: POLR3GL (polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse