Order Entry
Sweden
ContactUsLinkComponent
Anti-ETV2 Rabbit Polyclonal Antibody [clone: 175CT3.2.5]
Anti-ETV2 Rabbit Polyclonal Antibody [clone: 175CT3.2.5]
Catalog # ABGEAP11311A
Supplier:  Abgent
Anti-ETV2 Rabbit Polyclonal Antibody [clone: 175CT3.2.5]
Catalog # ABGEAP11311A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ETS variant 2
  • Antigen symbol:
    ETV2
  • Clonality:
    Polyclonal
  • Clone:
    175CT3.2.5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Ets-related protein 71|ER71|ETSRP71|Ets related protein 71|ETS translocation variant 2|Ets variant gene 2|ETV2_HUMAN.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ETV2 (ets variant 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human