Order Entry
Sweden
ContactUsLinkComponent
Anti-GFER Rabbit Polyclonal Antibody [clone: 21CT54.13.1 / 21CT54.13.7]
Anti-GFER Rabbit Polyclonal Antibody [clone: 21CT54.13.1 / 21CT54.13.7]
  ABGEAP11198C
 :  Abgent
Anti-GFER Rabbit Polyclonal Antibody [clone: 21CT54.13.1 / 21CT54.13.7]
  ABGEAP11198C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Growth factor, augmenter of liver regeneration
  • Antigen symbol:
    GFER
  • Clonality:
    Polyclonal
  • Clone:
    21CT54.13.1 / 21CT54.13.7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    HPO1|HPO2|HPO|HERV1|HSS|ALR|ERV1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: GFER (growth factor, augmenter of liver regeneration)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse