Order Entry
Sweden
ContactUsLinkComponent
Anti-MASTL Mouse Monoclonal Antibody
Anti-MASTL Mouse Monoclonal Antibody
Catalog # ABGEAM1911B
Supplier:  Abgent
undefined
Anti-MASTL Mouse Monoclonal Antibody
Catalog # ABGEAM1911B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    microtubule associated serine/threonine kinase-like
  • Antigen symbol:
    MASTL
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MASTL (microtubule associated serine/threonine kinase-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human