Anti-RGS3 Rabbit Polyclonal Antibody
Catalog # AVIVAVARP09007P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Regulator of G-protein signaling 3
- Antigen symbol:RGS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5998
- Antigen synonyms:C2PA|RGP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RGS3 (regulator of G-protein signaling 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: