Anti-USP17L3 Rabbit Polyclonal Antibody
Catalog # AVIVARP71944_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 17-like family member 3
- Antigen symbol:USP17L3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:645836
- Antigen synonyms:USP17F|USP17B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: USP17L3 (ubiquitin specific peptidase 17-like family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human