Anti-OR2A7 Rabbit Polyclonal Antibody
Catalog # AVIVARP71009_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:olfactory receptor, family 2, subfamily A, member 7
- Antigen symbol:OR2A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:401427
- Antigen synonyms:HSDJ0798C17|OR2A21
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: OR2A7 (olfactory receptor, family 2, subfamily A, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human