Anti-ANKRD40 Rabbit Polyclonal Antibody
AVIVARP69113_P050
- Antikroppstyp:Primär
- Antikroppsnamn:Ankyrin repeat domain 40
- Antigensymbol:ANKRD40
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:91369
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: ANKRD40 (ankyrin repeat domain 40)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human