Anti-GAGE12I Rabbit Polyclonal Antibody
Catalog # AVIVARP68370_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:G antigen 12I
- Antigen symbol:GAGE12I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:26748
- Antigen synonyms:AL4|CT4.7|GAGE-7B|GAGE7B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GAGE12I (G antigen 12I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human