Anti-ANAPC15 Rabbit Polyclonal Antibody
AVIVARP68090_P050
- Antibody type:Primary
- Antigen name:anaphase promoting complex subunit 15
- Antigen symbol:ANAPC15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25906
- Antigen synonyms:APC15|C11orf51|HSPC020
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ANAPC15 (anaphase promoting complex subunit 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human