Anti-EFNA5 Rabbit Polyclonal Antibody
Catalog # AVIVARP65431_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ephrin-A5
- Antigen symbol:EFNA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1946
- Antigen synonyms:AF1|EFL5|LERK7|RAGS|EPLG7|GLC1M
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EFNA5 (ephrin-A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human