Anti-COQ7 Rabbit Polyclonal Antibody
Catalog # AVIVARP64009_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:coenzyme Q7 homolog, ubiquinone (yeast)
- Antigen symbol:COQ7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10229
- Antigen synonyms:CLK-1|CAT5|CLK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: COQ7 (coenzyme Q7 homolog, ubiquinone (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human