Anti-DRD3 Rabbit Polyclonal Antibody
Catalog # AVIVARP63162_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Dopamine receptor D3
- Antigen symbol:DRD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1814
- Antigen synonyms:ETM1|FET1|D3DR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DRD3 (dopamine receptor D3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: