Anti-MRPS31 Rabbit Polyclonal Antibody
AVIVARP62691_P050
- Antibody type:Primary
- Antigen name:mitochondrial ribosomal protein S31
- Antigen symbol:MRPS31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10240
- Antigen synonyms:MRP-S31|S31mt|IMOGN38
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MRPS31 (mitochondrial ribosomal protein S31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human