Anti-NHEJ1 Rabbit Polyclonal Antibody
AVIVARP61619_P050
- Antibody type:Primary
- Antigen name:Nonhomologous end-joining factor 1
- Antigen symbol:NHEJ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:79840
- Antigen synonyms:XLF
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NHEJ1 (nonhomologous end-joining factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: