Order Entry
Sweden
ContactUsLinkComponent
Anti-ATCAY Rabbit Polyclonal Antibody
Anti-ATCAY Rabbit Polyclonal Antibody
Catalog # AVIVARP60863_P050
undefined
Anti-ATCAY Rabbit Polyclonal Antibody
Catalog # AVIVARP60863_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ataxia, cerebellar, Cayman type
  • Antigen symbol:
    ATCAY
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    85300
  • Antigen synonyms:
    BNIP-H|CLAC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: ATCAY (ataxia, cerebellar, Cayman type)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: