Anti-BSND Rabbit Polyclonal Antibody
Catalog # AVIVARP59578_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:barttin CLCNK-type chloride channel accessory beta subunit
- Antigen symbol:BSND
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7809
- Antigen synonyms:DFNB73|BART
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: BSND (Bartter syndrome, infantile, with sensorineural deafness (Barttin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human