Anti-TUB Rabbit Polyclonal Antibody
Catalog # AVIVARP57909_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tubby bipartite transcription factor
- Antigen symbol:TUB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:7275
- Antigen synonyms:rd5|RDOB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TUB (tubby bipartite transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: