Anti-RAB5C Rabbit Polyclonal Antibody
Catalog # AVIVARP57818_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: RAB5C (RAB5C, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Documents
ARP57818_P050.pdf
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:middle
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:RAB5C, member RAS oncogene family
- Cat. No.:AVIVARP57818_P050
- Gene ID:5878
- Antigen synonyms:RAB5CL|L1880|RAB5L|RABL
- Conjugation:Unconjugated
- Antigen symbol:RAB5C



