Anti-PRCC Rabbit Polyclonal Antibody
AVIVARP57814_P050
- Antibody type:Primary
- Antigen name:papillary renal cell carcinoma (translocation-associated)
- Antigen symbol:PRCC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:310687
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PRCC (papillary renal cell carcinoma (translocation-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat