Anti-TLDC1 Rabbit Polyclonal Antibody
AVIVARP57503_P050
- Antibody type:Primary
- Antigen name:TBC/LysM-associated domain containing 1
- Antigen symbol:TLDC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57707
- Antigen synonyms:KIAA1609
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TLDC1 (TBC/LysM-associated domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human