Anti-PLS3 Rabbit Polyclonal Antibody
Katalog # AVIVARP56623_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:Plastin 3
- Antigen symbol:PLS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:5358
- Antigen synonyms:BMND18|T-plastin
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: PLS3 (plastin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: