Anti-BEND7 Rabbit Polyclonal Antibody
Catalog # AVIVARP55523_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:BEN domain containing 7
- Antigensymbol:BEND7
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:222389
- Antigen synonymer:C10orf30
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: BEND7 (BEN domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human