Anti-PYCR2 Rabbit Polyclonal Antibody
Catalog # AVIVARP54937_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Pyrroline-5-carboxylate reductase 2
- Antigen symbol:PYCR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:29920
- Antigen synonyms:HLD10|P5CR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PYCR2 (pyrroline-5-carboxylate reductase family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: