Anti-COPG2 Rabbit Polyclonal Antibody
Catalog # AVIVARP54845_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:coatomer protein complex, subunit gamma 2
- Antigen symbol:COPG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:26958
- Antigen synonyms:2-COP|gamma-2-COP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: COPG2 (coatomer protein complex, subunit gamma 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: