Anti-2410015M20Rik Rabbit Polyclonal Antibody
Catalog # AVIVARP53503_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:RIKEN cDNA 2410015M20 gene
- Antigen symbol:2410015M20RIK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:224904
- Antigen synonyms:QIL1|sr104|Mic13
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: 2410015M20Rik
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: