Anti-CHMP4B Rabbit Polyclonal Antibody
Catalog # AVIVARP53339_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Charged multivesicular body protein 4B
- Antigen symbol:CHMP4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:128866
- Antigen synonyms:CTRCT31|CHMP4A|SNF7-2|C20orf178|Shax1|dJ553F4.4|VPS32B|SNF7|CTPP3|Vps32-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CHMP4B (charged multivesicular body protein 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: