Anti-PCP2 Rabbit Polyclonal Antibody
AVIVARP53294_P050
- Antibody type:Primary
- Antigen name:Purkinje cell protein 2
- Antigen symbol:PCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:126006
- Antigen synonyms:GPSM4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PCP2 (Purkinje cell protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: