Anti-RHPN1 Rabbit Polyclonal Antibody
AVIVARP52450_P050
- Antibody type:Primary
- Antigen name:Rhophilin Rho GTPase Binding Protein 1
- Antigen symbol:RHPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:114822
- Antigen synonyms:RHOPHILIN|RHPN|ODF5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RHPN1 (rhophilin, Rho GTPase binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: