Anti-TUBB4A Rabbit Polyclonal Antibody
AVIVARP52132_P050
- Antibody type:Primary
- Antigen name:tubulin, beta 4A class IVa
- Antigen symbol:TUBB4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10382
- Antigen synonyms:beta-5|TUBB4|DYT4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TUBB4A (tubulin, beta 4A class IVa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: