Order Entry
Sweden
ContactUsLinkComponent
Anti-DNAJA2 Rabbit Polyclonal Antibody
Anti-DNAJA2 Rabbit Polyclonal Antibody
  AVIVARP52128_P050
Anti-DNAJA2 Rabbit Polyclonal Antibody
  AVIVARP52128_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    DnaJ (Hsp40) homolog, subfamily A, member 2
  • Antigen symbol:
    DNAJA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    10294
  • Antigen synonyms:
    HIRIP4|PRO3015|DNAJ|RDJ2|DNJ3|DJA2|DJ3|CPR3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: DNAJA2 (DnaJ (Hsp40) homolog, subfamily A, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: