Anti-TRIM66 Rabbit Polyclonal Antibody
Catalog # AVIVARP51736_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 66
- Antigen symbol:TRIM66
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:9866
- Antigen synonyms:TIF1DELTA|C11orf29|TIF1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TRIM66 (tripartite motif containing 66)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: