Anti-TCEAL7 Rabbit Polyclonal Antibody
Catalog # AVIVARP50797_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transcription elongation factor A (SII)-like 7
- Antigen symbol:TCEAL7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Bovine
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:100040972
- Antigen synonyms:1110018P05Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TCEAL7 (transcription elongation factor A (SII)-like 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine