Order Entry
Sweden
ContactUsLinkComponent
Anti-GDPD5 Rabbit Polyclonal Antibody
Anti-GDPD5 Rabbit Polyclonal Antibody
Catalog # AVIVARP49968_P050
Anti-GDPD5 Rabbit Polyclonal Antibody
Catalog # AVIVARP49968_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycerophosphodiester phosphodiesterase domain containing 5
  • Antigen symbol:
    GDPD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    81544
  • Antigen synonyms:
    GDE2|PP1665
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: GDPD5 (glycerophosphodiester phosphodiesterase domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: