Anti-HM13 Rabbit Polyclonal Antibody
AVIVARP49964_P050
- Antibody type:Primary
- Antigen name:histocompatibility 13
- Antigen symbol:H13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:14950
- Antigen synonyms:5031424B04Rik|1200006O09Rik|AV020344|H-13|PSL3|Spp|Hm13|4930443L17Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HM13 (histocompatibility (minor) 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse