Anti-TMEM38A Rabbit Polyclonal Antibody
AVIVARP49799_P050
- Antibody type:Primary
- Antigen name:transmembrane protein 38A
- Antigen symbol:TMEM38A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79041
- Antigen synonyms:TRIC-A|TRICA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TMEM38A (transmembrane protein 38A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: