Anti-CYP4F12 Rabbit Polyclonal Antibody
Catalog # AVIVARP49789_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 4, subfamily F, polypeptide 12
- Antigen symbol:CYP4F12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:66002
- Antigen synonyms:CYPIVF12|F22329_1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CYP4F12 (cytochrome P450, family 4, subfamily F, polypeptide 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human