Anti-PORCN Rabbit Polyclonal Antibody
Catalog # AVIVARP49763_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:porcupine homolog (Drosophila)
- Antigen symbol:PORCN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:53627
- Antigen synonyms:AW045557|Mporc|Ppn|2410004O13Rik|mMg61|porc|Mg61|DXHXS7465e
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PORCN (porcupine homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: