Anti-TCTA Rabbit Polyclonal Antibody
AVIVARP49704_P050
- Antibody type:Primary
- Antigen name:T-cell leukemia translocation altered
- Antigen symbol:TCTA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6988
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TCTA (T-cell leukemia translocation altered)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat