Anti-LIPT2 Rabbit Polyclonal Antibody
Catalog # AVIVARP49255_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:lipoyl(octanoyl) transferase 2 (putative)
- Antigen symbol:LIPT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:387787
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LIPT2 (lipoyl(octanoyl) transferase 2 (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human