Anti-ST6GALNAC3 Rabbit Polyclonal Antibody
Katalog # AVIVARP49075_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3
- Antigen symbol:ST6GALNAC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:256435
- Antigen synonyms:ST6GALNACIII|PRO7177|STY|SIAT7C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: ST6GALNAC3 (ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: