Anti-TMTC2 Rabbit Polyclonal Antibody
Catalog # AVIVARP49064_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transmembrane and tetratricopeptide repeat containing 2
- Antigen symbol:TMTC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:160335
- Antigen synonyms:IBDBP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TMTC2 (transmembrane and tetratricopeptide repeat containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: