Anti-PNPLA5 Rabbit Polyclonal Antibody
Catalog # AVIVARP49010_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:patatin-like phospholipase domain containing 5
- Antigen symbol:PNPLA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:150379
- Antigen synonyms:dJ388M5.4|GS2L|dJ388M5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PNPLA5 (patatin-like phospholipase domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human