Anti-STK39 Rabbit Polyclonal Antibody
Katalog # AVIVARP48744_T100
Leverantör: Aviva Systems Biology
Specifikationer
- Antibody type:Primary
- Antigen name:Serine Threonine Kinase 39
- Antigen symbol:STK39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:27347
- Antigen synonyms:PASK|SPAK|DCHT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: