Anti-GGPS1 Rabbit Polyclonal Antibody
AVIVARP48534_P050
- Antibody type:Primary
- Antigen name:Geranylgeranyl diphosphate synthase 1
- Antigen symbol:GGPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:9453
- Antigen synonyms:GGPPS1|GGPPS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GGPS1 (geranylgeranyl diphosphate synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: